Gene Transcripts

Gene Symbol
Svs3b
Entrez Gene ID
329557
Gene Name
seminal vesicle secretory protein 3B
Gene Aliases
9530004A22Rik
Chromosome Location
Chr. 2: 164254363 - 164256643 on Build GRCm38
UniGene ID
Mm.118769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173377.2 NP_775553.1 3 1167
GenBank mRNA AK035235.1 - 3 1164

View Full Product Details