Gene Transcripts

Gene Symbol
Lyrm4
Entrez Gene ID
380840
Gene Name
LYR motif containing 4
Gene Aliases
BC034664, Gm903
Chromosome Location
Chr. 13: 35978797 - 36117357 on Build GRCm38
UniGene ID
Mm.390576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201358.2 NP_958746.1 3 431
GenBank mRNA AK050553.1 - 3 406
BC034664.1 - 3 329 5355517
BC080772.1 - 3 306 30139193

View Full Product Details