Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252347.1 | NP_001239276.1 | 6 | 890 | |
NM_016759.4 | NP_058039.1 | 6 | 890 | ||
GenBank mRNA | AK002856.1 | - | 2 | 188 | |
BC022745.1 | - | 4 | 615 | ||
BC046319.1 | - | 6 | 790 | 6391042 | |
U73941.1 | - | 6 | 617 |