Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020587.2 | NP_065612.2 | 4, 5 | 712 | |
GenBank mRNA | AB041587.1 | - | 1, 2 | 150 | |
AK075769.1 | - | 4, 5 | 680 | ||
AK077648.1 | - | 4, 5 | 698 | ||
AK156989.1 | - | 4, 5 | 647 | ||
BC019437.1 | - | 4, 5 | 672 | 4018178 | |
BC026944.1 | - | 4, 5 | 1578 | 4016087 | |
BC040812.1 | - | 4, 5 | 1578 |