Gene Transcripts

Gene Symbol
Mospd3
Entrez Gene ID
68929
Gene Name
motile sperm domain containing 3
Gene Aliases
1190005J19Rik, 5133401H10Rik, Gtig2, R124
Chromosome Location
Chr. 5: 137596645 - 137601083 on Build GRCm38
UniGene ID
Mm.41355
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254762.1 NP_001241691.1 4 891
NM_030037.2 NP_084313.1 5 1044
GenBank mRNA AK088731.1 - 4 882
BC003880.1 - 4 901 3496491
BC034167.1 - 5 1029
BC039620.1 - 5 1025 5051528
BQ445267.1 - 3 457
DV656015.1 - 2 301

View Full Product Details