Gene Transcripts

Gene Symbol
C8g
Entrez Gene ID
69379
Gene Name
complement component 8, gamma polypeptide
Gene Aliases
Chromosome Location
Chr. 2: 25498650 - 25501926 on Build GRCm38
UniGene ID
Mm.24596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027062.2 NP_081338.1 1, 2 413
GenBank mRNA BC019967.1 - 1, 2 413
BY090016.1 - 1, 2 182

View Full Product Details