Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006555.3 | NP_006546.1 | 2 | 293 | |
XM_005249582.4 | XP_005249639.1 | 2 | 294 | ||
XM_017011682.1 | XP_016867171.1 | 2 | 304 | ||
GenBank mRNA | AB451317.1 | - | 2 | 136 | |
AB451447.1 | - | 2 | 136 | ||
AF091079.1 | - | 2 | 196 | ||
AK223488.1 | - | 2 | 250 | ||
AK299158.1 | - | 2 | 304 | ||
AK312622.1 | - | 2 | 282 | ||
AU130339.1 | - | 2 | 304 | ||
BC007319.2 | - | 2 | 223 | 2989381 | |
BT007078.1 | - | 2 | 136 | ||
CR456989.1 | - | 2 | 136 | ||
CR542008.1 | - | 2 | 136 | ||
U95735.1 | - | 2 | 195 |