Gene Transcripts

Gene Symbol
RAI2
Entrez Gene ID
10742
Gene Name
retinoic acid induced 2
Gene Aliases
Chromosome Location
Chr. X: 17800049 - 17861346 on Build GRCh38
UniGene ID
Hs.446680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172732.1 NP_001166203.1 3 1619
NM_001172739.1 NP_001166210.1 3 1963
NM_001172743.1 NP_001166214.1 3 1966
NM_021785.4 NP_068557.3 2 1769
NR_033348.1 3 2042
NR_033349.1 2 1848
XM_006724459.2 XP_006724522.1 4 2118
XM_006724460.1 XP_006724523.1 3 1866
XM_011545439.2 XP_011543741.1 4 2147
XM_011545440.2 XP_011543742.1 4 2150
XM_011545441.2 XP_011543743.1 3 1953
GenBank mRNA AK056214.1 - 3 1862
AK298873.1 - 3 1580
AK314631.1 - 2 1732
BC027937.1 - 3 1922 5201821

View Full Product Details