Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145158.1 | NP_001138630.1 | 12 | 1631 | |
NM_006620.3 | NP_006611.1 | 13 | 1757 | ||
GenBank mRNA | AB028961.1 | - | 8 | 986 | |
AJ459826.1 | - | 13 | 1743 | ||
AK024258.1 | - | 13 | 1678 | ||
AK293573.1 | - | 10 | 965 | ||
AK295545.1 | - | 12 | 1552 | ||
AK298336.1 | - | 10 | 1160 | ||
AL137664.1 | - | 1 | 1069 | ||
BC001465.2 | - | 13 | 1666 | 3138910 | |
BC040849.1 | - | 13 | 1661 | 5104748 | |
BX648033.1 | - | 10 | 1422 | ||
U87791.1 | - | 13 | 1744 |