Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191005.2 | NP_001177934.1 | 3 | 393 | |
NM_001191006.2 | NP_001177935.1 | 3 | 393 | ||
NM_001191007.2 | NP_001177936.1 | 3 | 393 | ||
NM_001191009.2 | NP_001177938.1 | 3 | 393 | ||
NM_001300936.1 | NP_001287865.1 | 3 | 393 | ||
NM_001300937.1 | NP_001287866.1 | 3 | 393 | ||
NM_006625.5 | NP_006616.1 | 3 | 393 | ||
NM_054016.3 | NP_473357.1 | 3 | 393 | ||
XM_017000100.1 | XP_016855589.1 | 3 | 4113 | ||
XM_017000101.1 | XP_016855590.1 | 4 | 3656 | ||
XM_017000102.1 | XP_016855591.1 | 4 | 1283 | ||
GenBank mRNA | AF047448.1 | - | 3 | 214 | |
AF067730.1 | - | 3 | 214 | ||
AF419331.1 | - | 3 | 261 | ||
AF449427.1 | - | 3 | 185 | ||
AK001286.1 | - | 3 | 257 | ||
AK001656.1 | - | 3 | 257 | ||
AK055311.1 | - | 1 | 1195 | ||
AK125834.1 | - | 1 | 2590 | ||
AK222994.1 | - | 3 | 284 | ||
AK296175.1 | - | 3 | 286 | ||
AY007101.1 | - | 3 | 226 | ||
AY150180.1 | - | 3 | 199 | ||
AY150181.1 | - | 3 | 199 | ||
BC001107.1 | - | 3 | 286 | 3504780 | |
BC005039.1 | - | 3 | 249 | 3531441 | |
BC010074.2 | - | 3 | 238 | 4302658 | |
BC071575.1 | - | 3 | 3514 | ||
BX648939.1 | - | 1 | 2446 | ||
KU178536.1 | - | 3 | 185 |