Gene Transcripts

Gene Symbol
Ostc
Entrez Gene ID
66357
Gene Name
oligosaccharyltransferase complex subunit
Gene Aliases
2310008M10Rik, 5730557H03Rik
Chromosome Location
Chr. 3: 130695917 - 130709444 on Build GRCm38
UniGene ID
Mm.300697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025509.3 NP_079785.1 1, 2 216
GenBank mRNA AK009237.1 - 1, 2 191
AK011085.1 - 1, 2 168
AK013011.1 - 1, 2 175
AK135332.1 - 1, 2 172
BC014799.1 - 1, 2 189
BC021935.1 - 1, 2 190 5149713
BY228396.1 - 1, 2 217

View Full Product Details