Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006658.4 | NP_006649.2 | 3 | 422 | |
XM_011515094.1 | XP_011513396.1 | 3 | 254 | ||
GenBank mRNA | AF071789.1 | - | 2 | 147 | |
AF097730.1 | - | 2 | 144 | ||
AK312989.1 | - | 3 | 224 | ||
BC028094.1 | - | 3 | 206 | 5241722 | |
DA061079.1 | - | 3 | 422 | ||
DB039499.1 | - | 3 | 254 |