Gene Transcripts

Gene Symbol
CCL27
Entrez Gene ID
10850
Gene Name
C-C motif chemokine ligand 27
Gene Aliases
ALP, CTACK, CTAK, ESKINE, ILC, PESKY, SCYA27
Chromosome Location
Chr. 9: 34661883 - 34662692 on Build GRCh38
UniGene ID
Hs.648124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006664.3 NP_006655.1 1 101
GenBank mRNA AB010445.1 - 1 101
AF082393.1 - 1 42
AJ243542.1 - 1 66
BC148263.1 - 1 64 8860203

View Full Product Details