Gene Transcripts

Gene Symbol
C1QL1
Entrez Gene ID
10882
Gene Name
complement C1q like 1
Gene Aliases
C1QRF, C1QTNF14, CRF
Chromosome Location
Chr. 17: 44959693 - 44968276 on Build GRCh38
UniGene ID
Hs.134012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006688.4 NP_006679.1 1 751
GenBank mRNA AF095154.1 - 1 536
BC008798.2 - 1 751 3635430

View Full Product Details