Gene Transcripts

Gene Symbol
ZPBP
Entrez Gene ID
11055
Gene Name
zona pellucida binding protein
Gene Aliases
ZPBP1
Chromosome Location
Chr. 7: 49846355 - 50093264 on Build GRCh38
UniGene ID
Hs.388841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159878.1 NP_001153350.1 4, 5 554
NM_007009.2 NP_008940.2 4, 5 557
XM_011515095.2 XP_011513397.1 4, 5 551
XM_011515096.2 XP_011513398.1 4, 5 526
XM_011515097.1 XP_011513399.1 4, 5 551
XM_011515100.2 XP_011513402.1 3, 4 379
XM_011515102.2 XP_011513404.1 4, 5 730
XM_011515103.1 XP_011513405.1 4, 5 551
XM_017011707.1 XP_016867196.1 4, 5 551
GenBank mRNA AK312799.1 - 4, 5 526
BC005223.1 - 4, 5 516 3950015
BT006822.1 - 4, 5 487
D17570.1 - 4, 5 523

View Full Product Details