Gene Transcripts
- Gene Symbol
- Gp6
- Entrez Gene ID
- 243816
- Gene Name
- glycoprotein 6 (platelet)
- Gene Aliases
- 9830166G18Rik, Gm469, Gpvi
- Chromosome Location
- Chr. 7: 4362853 - 4397905 on Build GRCm38
- UniGene ID
- Mm.248662
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC138170.1 |
- |
4 |
505 |
|
BC145172.1 |
- |
4 |
505 |
9053723 |
View Full Product Details