Gene Transcripts
- Gene Symbol
- Crygf
- Entrez Gene ID
- 689947
- Gene Name
- crystallin, gamma F
- Gene Aliases
- Cryg6, Len
- Chromosome Location
- Chr. 9: 72933304 - 72935468 on Build Rnor_6.0
- UniGene ID
- Rn.102739
- Species
- Rattus norvegicus
- Gene Symbol
- Crygd
- Entrez Gene ID
- 24278
- Gene Name
- crystallin, gamma D
- Gene Aliases
- Cryg4, Len
- Chromosome Location
- Chr. 9: 71776565 - 71779107 on Build Rnor_6.0
- UniGene ID
- Rn.64655
- Species
- Rattus norvegicus
- Gene Symbol
- Cryge
- Entrez Gene ID
- 24279
- Gene Name
- crystallin, gamma E
- Gene Aliases
- Cryg5, Len
- Chromosome Location
- Chr. 9: 71764317 - 71767411 on Build Rnor_6.0
- UniGene ID
- Rn.44578
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173289.1 |
NP_775411.1
|
2 |
188 |
|
GenBank mRNA |
J00716.1 |
- |
|
146 |
|
View Full Product Details