Gene Transcripts

Gene Symbol
Bnip1
Entrez Gene ID
140932
Gene Name
BCL2/adenovirus E1B interacting protein 1
Gene Aliases
Bnip1l1
Chromosome Location
Chr. 10: 16677077 - 16689321 on Build Rnor_6.0
UniGene ID
Rn.16757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080897.1 NP_543173.1 6 533
GenBank mRNA AF454391.1 - 6 533
FQ218385.1 - 6 537

View Full Product Details