Gene Transcripts

Gene Symbol
BAZ1A
Entrez Gene ID
11177
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
ACF1, WALp1, WCRF180, hACF1
Chromosome Location
Chr. 14: 34752731 - 34875647 on Build GRCh38
UniGene ID
Hs.509140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013448.2 NP_038476.2 3 915
NM_182648.1 NP_872589.1 3 915
GenBank mRNA AB032252.1 - 3 912
AF161435.1 - 3 601
AF213467.1 - 3 513
BC020636.1 - 2 675
BC108711.1 - 3 579

View Full Product Details