Gene Transcripts

Gene Symbol
Snx15
Entrez Gene ID
293691
Gene Name
sorting nexin 15
Gene Aliases
Chromosome Location
Chr. 1: 221487058 - 221496200 on Build Rnor_6.0
UniGene ID
Rn.104898
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024752.1 NP_001019923.1 2, 3 232
GenBank mRNA BC097481.1 - 2, 3 232

View Full Product Details