Gene Transcripts

Gene Symbol
INMT
Entrez Gene ID
11185
Gene Name
indolethylamine N-methyltransferase
Gene Aliases
TEMT
Chromosome Location
Chr. 7: 30752135 - 30757602 on Build GRCh38
UniGene ID
Hs.632629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199219.1 NP_001186148.1 3 530
NM_006774.4 NP_006765.4 3 533
GenBank mRNA AF128846.1 - 3 533
AF128847.1 - 3 533
AK309519.1 - 4 663
AK313832.1 - 3 528
BC033813.1 - 3 515
BC103712.1 - 3 516
BC106902.1 - 3 533 40031569
BC106903.1 - 3 533 40031570
DA920005.1 - 3 533

View Full Product Details