Gene Transcripts

Gene Symbol
Hnrnpa3
Entrez Gene ID
362152
Gene Name
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases
Hnrpa3
Chromosome Location
Chr. 3: 62481228 - 62491356 on Build Rnor_6.0
UniGene ID
Rn.107690
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111294.1 NP_001104764.1 6 760
NM_001111295.1 NP_001104765.1 6 694
NM_198132.3 NP_937765.1 6 760
XM_008761911.1 XP_008760133.1 6 709
XM_008761912.1 XP_008760134.1 6 709
XM_008761913.1 XP_008760135.1 6 755
GenBank mRNA AY363226.1 - 6 706
AY363227.1 - 6 640
BC081878.1 - 6 700 7108128
CB725005.1 - 6 632
FQ214617.1 - 6 656
Y16641.1 - 2 122

View Full Product Details