Gene Transcripts

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
Chromosome Location
Chr. 3: 99833144 - 100114513 on Build GRCh38
UniGene ID
Hs.104672
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042459.2 NP_001035924.1 5 1475
NM_001282794.1 NP_001269723.1 2 531
NM_014890.3 NP_055705.2 2 531
NM_182909.3 NP_878913.2 5 1475
XM_006713486.2 XP_006713549.1 3 727
XM_011512371.2 XP_011510673.1 3 593
GenBank mRNA AF329092.1 - 5 1476
AF514867.1 - 4 1004
AF514868.1 - 4 1004
AF514869.1 - 4 1004
AK096195.1 - 3 326
AK225855.1 - 3 671
AX776292.1 - 5 1476
AX776294.1 - 4 1012
AX776296.1 - 4 1015
AX776298.1 - 4 1012
AY642382.1 - 4 1019
BC027860.1 - 2 462 5228947
BP233250.1 - 2 530
EU531865.1 - 2 432
U53445.1 - 1 418

View Full Product Details