Gene Transcripts

Gene Symbol
Luc7l3
Entrez Gene ID
360602
Gene Name
LUC7-like 3 (S. cerevisiae)
Gene Aliases
Crop, RGD1307981
Chromosome Location
Chr. 10: 81991147 - 82027119 on Build Rnor_6.0
UniGene ID
Rn.233772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247156.2 XP_006247218.1 10 1448
XM_008768078.1 XP_008766300.1 10 1448
GenBank mRNA FQ212347.1 - 10 1428
FQ235260.1 - 3 471

View Full Product Details