Gene Transcripts

Gene Symbol
U2AF2
Entrez Gene ID
11338
Gene Name
U2 small nuclear RNA auxiliary factor 2
Gene Aliases
U2AF65
Chromosome Location
Chr. 19: 55654050 - 55674716 on Build GRCh38
UniGene ID
Hs.528007
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012478.1 NP_001012496.1 6 1642
NM_007279.2 NP_009210.1 6 1642
XM_006722994.2 XP_006723057.1 8 1740
XM_011526410.1 XP_011524712.1 7 1680
GenBank mRNA AB451261.1 - 6 587
AB451387.1 - 6 587
BC008740.2 - 6 646 3346087
BC030574.2 - 6 1546 3687471
X64044.1 - 6 627

View Full Product Details