Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052847.2 | NP_443079.1 | 5 | 739 | |
XM_017026606.1 | XP_016882095.1 | 5 | 848 | ||
GenBank mRNA | AB010414.1 | - | 3 | 579 | |
BC014466.1 | - | 5 | 698 | 4861393 | |
BC053630.1 | - | 5 | 654 | 6469872 |