Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113546.1 | NP_001107018.1 | 11 | 2207 | |
NM_001113547.1 | NP_001107019.1 | 8 | 2088 | ||
NM_001243775.1 | NP_001230704.1 | 5 | 1815 | ||
NM_016357.4 | NP_057441.1 | 11 | 2204 | ||
XM_011538455.2 | XP_011536757.1 | 11 | 2267 | ||
XM_017019429.1 | XP_016874918.1 | 6 | 1717 | ||
XM_017019430.1 | XP_016874919.1 | 11 | 2558 | ||
GenBank mRNA | AB209512.1 | - | 11 | 2143 | |
AF157325.1 | - | 5 | 1814 | ||
AF198454.1 | - | 11 | 2131 | ||
AF198455.1 | - | 8 | 2026 | ||
AF218025.1 | - | 7 | 1338 | ||
AK000057.1 | - | 5 | 1130 | ||
AK000372.1 | - | 11 | 2114 | ||
AK023649.1 | - | 5 | 1803 | ||
AK096172.1 | - | 5 | 2395 | ||
AK222971.1 | - | 11 | 2127 | ||
AK297766.1 | - | 9 | 1800 | ||
AK314447.1 | - | 11 | 2116 | ||
AL136911.1 | - | 11 | 2123 | ||
BC001247.1 | - | 8 | 1733 | ||
BC010664.1 | - | 1 | 581 | ||
BC110815.1 | - | 11 | 2141 | 5115521 | |
BC136763.1 | - | 11 | 2090 | 9020753 | |
BC144402.1 | - | 12 | 2146 | ||
BC144403.1 | - | 12 | 2149 |