Gene Transcripts

Gene Symbol
TSHB
Entrez Gene ID
7252
Gene Name
thyroid stimulating hormone beta
Gene Aliases
TSH-B, TSH-BETA
Chromosome Location
Chr. 1: 115029824 - 115035935 on Build GRCh38
UniGene ID
Hs.406687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000549.4 NP_000540.2 2 148
XM_011542065.2 XP_011540367.1 1 187
GenBank mRNA BC069298.1 - 1 110

View Full Product Details