Gene Transcripts

Gene Symbol
NLRP1
Entrez Gene ID
22861
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1
Chromosome Location
Chr. 17: 5501399 - 5584512 on Build GRCh38
UniGene ID
Hs.652273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033053.2 NP_001028225.1 4 2729
NM_014922.4 NP_055737.1 4 2729
NM_033004.3 NP_127497.1 4 2729
NM_033006.3 NP_127499.1 4 2729
NM_033007.3 NP_127500.1 4 2729
GenBank mRNA AB023143.1 - 4 2696
AF229059.1 - 4 2174
AF229060.1 - 4 2174
AF229061.1 - 4 2174
AF229062.1 - 4 2174
AF310105.1 - 4 2692
AK026393.1 - 1 652
AK310936.1 - 5 2540
AL117470.1 - 1 1332

View Full Product Details