Gene Transcripts

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
Chromosome Location
Chr. 14: 55366391 - 55411858 on Build GRCh38
UniGene ID
Hs.414809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014924.4 NP_055739.2 1, 2 259
GenBank mRNA AK131251.1 - 1, 2 259
AK292810.1 - 1, 2 231
BC109089.1 - 1, 2 244 40034573
BC109090.1 - 1, 2 244 40034574

View Full Product Details