Gene Transcripts

Gene Symbol
CA5A
Entrez Gene ID
763
Gene Name
carbonic anhydrase 5A
Gene Aliases
CA5, CA5AD, CAV, CAVA, GS1-21A4.1
Chromosome Location
Chr. 16: 87888019 - 87936575 on Build GRCh38
UniGene ID
Hs.177446
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001739.1 NP_001730.1 3, 4 526
XM_011523309.2 XP_011521611.1 3, 4 595
XM_011523310.2 XP_011521612.1 3, 4 595
XM_017023646.1 XP_016879135.1 3, 4 595
GenBank mRNA BC137405.1 - 3, 4 522
BC137411.1 - 3, 4 522 9021409
L19297.1 - 3, 4 525

View Full Product Details