Gene Transcripts

Gene Symbol
IZUMO1
Entrez Gene ID
284359
Gene Name
izumo sperm-egg fusion 1
Gene Aliases
IZUMO, OBF
Chromosome Location
Chr. 19: 48740888 - 48747499 on Build GRCh38
UniGene ID
Hs.400688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321864.1 NP_001308793.1 2 231
NM_001321865.1 NP_001308794.1 2 357
NM_182575.2 NP_872381.2 3 832
NR_135832.1 2 290
XM_005258793.4 XP_005258850.1 3 565
XM_011526811.2 XP_011525113.1 2 1336
XM_011526812.2 XP_011525114.1 3 912
XM_011526813.2 XP_011525115.1 3 761
XM_011526815.2 XP_011525117.1 2 1475
XM_017026646.1 XP_016882135.1 2 1336
GenBank mRNA AB195682.1 - 3 690
AK301750.1 - 2 1475
AY552609.1 - 2 284
AY552610.1 - 2 217
BC034769.1 - 3 832 5167853

View Full Product Details