Gene Transcripts

Gene Symbol
GOLGA6L9
Entrez Gene ID
440295
Gene Name
golgin A6 family-like 9
Gene Aliases
GOLGA6L20
Chromosome Location
Chr. 15: 82429363 - 82439387 on Build GRCh38
UniGene ID
Hs.498322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291420.1 NP_001278349.1 8 1362
NM_198181.3 NP_937824.3 9 1494
XM_011521601.2 XP_011519903.1 8 2045
XM_017022226.1 XP_016877715.1 7 1481
GenBank mRNA AK128268.1 - 5 1041
BC041854.1 - 8 1363 5270411
CR936812.1 - 6 3897
DR157204.1 - 1 238

View Full Product Details