Gene Transcripts

Gene Symbol
RPS14
Entrez Gene ID
6208
Gene Name
ribosomal protein S14
Gene Aliases
EMTB, S14
Chromosome Location
Chr. 5: 150444229 - 150449810 on Build GRCh38
UniGene ID
Hs.744846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025070.1 NP_001020241.1 5 733
NM_001025071.1 NP_001020242.1 5 739
NM_005617.3 NP_005608.1 5 522
GenBank mRNA AA467888.1 - 3 289
BC001126.1 - 1 485 2967098
BC003401.1 - 4 479 3448752
BC006784.1 - 1 490 2900907
BC020515.1 - 5 672 3856643
BC091474.1 - 5 503 30334486
BM454896.1 - 5 702
BM557014.1 - 5 674

Gene Symbol
RPS14P3
Entrez Gene ID
644068
Gene Name
ribosomal protein S14 pseudogene 3
Gene Aliases
RPS14_1_16
Chromosome Location
Chr. 1: 19607807 - 19608644 on Build GRCh38
UniGene ID
Hs.660760
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_077246.1 1 772
GenBank mRNA BC072682.1 - 1 772

View Full Product Details