Gene Transcripts

Gene Symbol
RPS20
Entrez Gene ID
6224
Gene Name
ribosomal protein S20
Gene Aliases
S20
Chromosome Location
Chr. 8: 56068180 - 56074581 on Build GRCh38
UniGene ID
Hs.8102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001023.3 NP_001014.1 2 261
NM_001146227.1 NP_001139699.1 2 261
GenBank mRNA AK130961.1 - 2 186
AK301342.1 - 2 190
AK311808.1 - 2 186
AV716603.1 - 190
BC007507.2 - 2 170 3029762
BC087850.1 - 2 156 30386230
BQ064297.1 - 2 171
CB138358.1 - 2 261
L06498.1 - 2 176

View Full Product Details