Gene Transcripts
- Gene Symbol
- RAB18
- Entrez Gene ID
- 22931
- Gene Name
- RAB18, member RAS oncogene family
- Gene Aliases
- RAB18LI1, WARBM3
- Chromosome Location
- Chr. 10: 27504174 - 27542237 on Build GRCh38
- UniGene ID
- Hs.406799
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256410.1 |
NP_001243339.1
|
6, 7 |
669 |
|
NM_001256415.1 |
NP_001243344.1
|
4, 5 |
510 |
|
NM_021252.4 |
NP_067075.1
|
5, 6 |
582 |
|
NR_046172.1 |
|
4, 5 |
520 |
|
GenBank mRNA |
AB209698.1 |
- |
1, 2 |
5344 |
|
AF087860.1 |
- |
5, 6 |
440 |
|
AF136974.1 |
- |
5, 6 |
496 |
|
AF137372.1 |
- |
5, 6 |
386 |
|
AF498950.1 |
- |
5, 6 |
386 |
|
AK001555.1 |
- |
5, 6 |
386 |
|
AK223153.1 |
- |
5, 6 |
409 |
|
AK299457.1 |
- |
4, 5 |
386 |
|
AL136734.1 |
- |
5, 6 |
435 |
|
AY574034.1 |
- |
6, 7 |
558 |
|
BC015014.2 |
- |
5, 6 |
436 |
3922029 |
BC029350.1 |
- |
5, 6 |
452 |
4819866 |
BP210053.1 |
- |
4, 5 |
384 |
|
BT009840.1 |
- |
5, 6 |
386 |
|
CN357871.1 |
- |
5, 6 |
651 |
|
CR533455.1 |
- |
5, 6 |
386 |
|
View Full Product Details