Gene Transcripts

Gene Symbol
PAXIP1
Entrez Gene ID
22976
Gene Name
PAX interacting protein 1
Gene Aliases
CAGF28, CAGF29, PACIP1, PAXIP1L, PTIP, TNRC2
Chromosome Location
Chr. 7: 154943690 - 155003393 on Build GRCh38
UniGene ID
Hs.443881
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007349.3 NP_031375.3 13, 14 2515
XM_005249539.1 XP_005249596.1 14, 15 2654
XM_011515982.2 XP_011514284.1 13, 14 2629
XM_011515983.1 XP_011514285.1 13, 14 2515
XM_017011881.1 XP_016867370.1 14, 15 3827
GenBank mRNA AK093584.1 - 7, 8 1208
AK123600.1 - 14, 15 2654
AK126674.1 - 5, 6 2523
AK293744.1 - 11, 12 2347
BC008328.1 - 4, 5 439
BC033781.1 - 8, 9 1756 4509264
BX538201.1 - 7, 8 1291
BX640616.1 - 13, 14 4413
U80735.1 - 9, 10 1498

View Full Product Details