Gene Transcripts

Gene Symbol
MRPL24
Entrez Gene ID
79590
Gene Name
mitochondrial ribosomal protein L24
Gene Aliases
L24mt, MRP-L18, MRP-L24
Chromosome Location
Chr. 1: 156737302 - 156741267 on Build GRCh38
UniGene ID
Hs.418233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024540.3 NP_078816.2 6 704
NM_145729.2 NP_663781.1 6 745
XM_011509981.2 XP_011508283.1 6 815
XM_011509982.2 XP_011508284.1 6 713
GenBank mRNA AK024570.1 - 6 702
AK222500.1 - 6 702
AK223066.1 - 6 702
BC012440.1 - 6 686 3863359
BC016700.1 - 6 704 4108596
BG706700.1 - 6 709
CR457329.1 - 5 565

View Full Product Details