Gene Transcripts

Gene Symbol
TMEM41A
Entrez Gene ID
90407
Gene Name
transmembrane protein 41A
Gene Aliases
2900010K02Rik
Chromosome Location
Chr. 3: 185482671 - 185499057 on Build GRCh38
UniGene ID
Hs.677570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080652.3 NP_542383.1 4 600
XM_017007437.1 XP_016862926.1 3 438
GenBank mRNA AK172748.1 - 3 1780
AK290878.1 - 4 578
AK313954.1 - 4 536
AL832782.1 - 1 59
AY358547.1 - 4 584
BC008043.2 - 4 557 3838280
BC019884.1 - 4 547 4991416
BX647128.1 - 2 250
BX647928.1 - 2 2472

View Full Product Details