Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000772.2 | NP_000763.1 | 8 | 1488 | |
NM_001128925.1 | NP_001122397.1 | 7 | 1311 | ||
GenBank mRNA | AK223510.1 | - | 8 | 1187 | |
AK308558.1 | - | 8 | 1143 | ||
AK313403.1 | - | 8 | 1187 | ||
BC069666.1 | - | 8 | 1356 | 7262569 | |
BC096257.1 | - | 8 | 1207 | 40006072 | |
BC096258.1 | - | 8 | 1207 | 40006073 | |
BC096260.1 | - | 7 | 1030 | 40006076 | |
BX538127.1 | - | 5 | 622 | ||
M61856.1 | - | 8 | 1361 |