Gene Transcripts

Gene Symbol
CLRN2
Entrez Gene ID
645104
Gene Name
clarin 2
Gene Aliases
Chromosome Location
Chr. 4: 17515165 - 17527104 on Build GRCh38
UniGene ID
Hs.631526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079827.2 NP_001073296.1 3 610
GenBank mRNA BC127863.1 - 3 610

View Full Product Details