Gene Transcripts

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
Chromosome Location
Chr. 11: 47565430 - 47573461 on Build GRCh38
UniGene ID
Hs.656205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143984.1 NP_001137456.1 2 614
NM_175732.2 NP_783859.1 4 697
GenBank mRNA AF277187.1 - 2 111
AK294640.1 - 3 336
AY333987.1 - 3 312
BC014048.1 - 3 343
BC020242.1 - 4 526 4591438
BC107573.1 - 2 449
BQ050653.1 - 2 90
DR004030.1 - 4 555

View Full Product Details