Gene Transcripts

Gene Symbol
SYCE1L
Entrez Gene ID
100130958
Gene Name
synaptonemal complex central element protein 1 like
Gene Aliases
MRP2
Chromosome Location
Chr. 16: 77199452 - 77213079 on Build GRCh38
UniGene ID
Hs.734779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129979.1 NP_001123451.1 6, 7 360

View Full Product Details