Gene Transcripts

Gene Symbol
DNAJC25-GNG10
Entrez Gene ID
552891
Gene Name
DNAJC25-GNG10 readthrough
Gene Aliases
Chromosome Location
Chr. 9: 111631352 - 111670246 on Build GRCh38
UniGene ID
Hs.534196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004125.3 NP_004116.2 1, 2 390
GenBank mRNA BC015391.1 - 1, 2 356
BT007068.1 - 1, 2 334

View Full Product Details