Gene Transcripts

Gene Symbol
MUSTN1
Entrez Gene ID
389125
Gene Name
musculoskeletal, embryonic nuclear protein 1
Gene Aliases
MUSTANG
Chromosome Location
Chr. 3: 52833115 - 52835219 on Build GRCh38
UniGene ID
Hs.556077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205853.3 NP_995325.3 3 685
GenBank mRNA AJ276556.1 - 3 530
AK310752.1 - 3 166
BX089772.1 - 3 670
CR990075.1 - 3 576

Gene Symbol
TMEM110-MUSTN1
Entrez Gene ID
100526772
Gene Name
TMEM110-MUSTN1 readthrough
Gene Aliases
Chromosome Location
Chr. 3: 52833115 - 52897581 on Build GRCh38
UniGene ID
Hs.744189
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198974.2 NP_001185903.2 10 1415
GenBank mRNA AL833659.1 - 4 5224

View Full Product Details