Gene Transcripts

Gene Symbol
Cyp2c68
Entrez Gene ID
433247
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 68
Gene Aliases
9030012A22Rik
Chromosome Location
Chr. 19: 39688836 - 39741101 on Build GRCm38
UniGene ID
Mm.335660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039555.2 NP_001034644.2 2 365
GenBank mRNA AI226521.1 - 2 366
BC019468.1 - 2 318 4160492

View Full Product Details