Gene Transcripts

Gene Symbol
Cyp2c68
Entrez Gene ID
433247
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 68
Gene Aliases
9030012A22Rik
Chromosome Location
Chr. 19: 39688836 - 39741101 on Build GRCm38
UniGene ID
Mm.335660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039555.2 NP_001034644.2 2 364
GenBank mRNA AI226521.1 - 2 365
BC019468.1 - 2 317 4160492

Gene Symbol
Cyp2c40
Entrez Gene ID
13099
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 40
Gene Aliases
AI265721, AI662255
Chromosome Location
Chr. 19: 39767073 - 39812814 on Build GRCm38
UniGene ID
Mm.353875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010004.2 NP_034134.2 2 396
GenBank mRNA AF047727.1 - 2 310
BC139471.1 - 2 396 8862493
BC139472.1 - 2 396 8862494

View Full Product Details