Gene Transcripts

Gene Symbol
RCOR1
Entrez Gene ID
23186
Gene Name
REST corepressor 1
Gene Aliases
COREST, RCOR
Chromosome Location
Chr. 14: 102592638 - 102730576 on Build GRCh38
UniGene ID
Hs.510521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015156.3 NP_055971.2 8 1265
GenBank mRNA AF155595.1 - 8 1028
D31888.1 - 8 770

View Full Product Details