Gene Transcripts

Gene Symbol
Ear6
Entrez Gene ID
93719
Gene Name
eosinophil-associated, ribonuclease A family, member 6
Gene Aliases
Chromosome Location
Chr. 14: 51853768 - 51854643 on Build GRCm38
UniGene ID
Mm.221324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053111.2 NP_444341.1 2 360
GenBank mRNA AK157836.1 - 2 357
AY316150.1 - 2 360
BC147690.1 - 1 592 9056663
BC147697.1 - 1 592 9056670

View Full Product Details