Gene Transcripts

Gene Symbol
Vmn2r13
Entrez Gene ID
231589
Gene Name
vomeronasal 2, receptor 13
Gene Aliases
EG231589, Gm4867
Chromosome Location
Chr. 5: 109156068 - 109192107 on Build GRCm38
UniGene ID
Mm.469883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104624.1 NP_001098094.1 2 379

View Full Product Details